Assay Details
Target Gene Details
Entrez Gene ID: | 54344 |
Gene Name: | dolichyl-phosphate mannosyltransferase subunit 3 |
Gene Aliases: |
CDG1O |
Location: |
Chr.1:155139891-155140573 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DPM3 | NM_018973.3 | 1 | 359 | NP_061846.2 |
| NM_153741.1 | 2 | 227 | NP_714963.1 | |
| XM_017001498.1 | 2 | 306 | XP_016856987.1 | |
| AB028128.1 | 1 | 209 | BAA96291.1 | |
| AF312922.1 | 1 | 359 | AAK28487.1 | |
| AF312923.1 | 1 | 209 | AAK28486.1 | |
| AK293625.1 | 1 | 483 | ||
| BC032223.2 | 1 | 198 | AAH32223.2 | |
| BC065233.1 | 2 | 235 | AAH65233.1 | |
| BC104202.1 | 1 | 332 | AAI04203.1 | |
| BC104203.2 | 1 | 332 | AAI04204.1 | |
| BC104481.1 | 2 | 255 | AAI04482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
| nsv547961 | Chr.1:155109822 - 155225189 on Build GRCh38 | Loss |
|
| nsv998756 | Chr.1:155107614 - 155164069 on Build GRCh38 | Gain |
|
| esv3587615 | Chr.1:155047853 - 155175013 on Build GRCh38 | Gain |
|
| dgv3n68 | Chr.1:155122502 - 155343618 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map