Assay Details
Target Gene Details
Entrez Gene ID: | 347527 |
Gene Name: | arylsulfatase family member H |
Gene Aliases: |
sulfatase |
Location: |
Chr.X:3006613-3033385 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSH | NM_001011719.1 | NP_001011719.1 | ||
| AY875940.1 | AAW66668.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33241 | Chr.X:2968057 - 3034162 on Build GRCh38 | Loss |
|
| esv34985 | Chr.X:2623614 - 3258451 on Build GRCh38 | Gain |
|
| esv2758855 | Chr.X:1703008 - 3276324 on Build GRCh38 | Gain |
|
| esv2674359 | Chr.X:2983683 - 3012017 on Build GRCh38 | Deletion |
|
| dgv16n29 | Chr.X:3010013 - 3194841 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs79487908] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map