Assay Details
Target Gene Details
Entrez Gene ID: | 51542 |
Gene Name: | VPS54, GARP complex subunit |
Gene Aliases: |
HCC8, PPP1R164, SLP-8p, VPS54L, WR, hVps54L |
Location: |
Chr.2:63892149-64019421 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 22 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS54 | NM_001005739.1 | NP_001005739.1 | ||
| NM_016516.2 | NP_057600.2 | |||
| XM_006712029.3 | XP_006712092.1 | |||
| XM_017004316.1 | XP_016859805.1 | |||
| AF102177.1 | AAF37319.1 | |||
| AK002205.1 | BAA92134.1 | |||
| AK025545.1 | ||||
| AK094019.1 | ||||
| AK124436.1 | ||||
| AK292669.1 | ||||
| AK292889.1 | ||||
| AL137604.1 | CAB70837.1 | |||
| AL359939.1 | CAB95772.1 | |||
| AY444798.1 | AAS20945.1 | |||
| BC030275.1 | AAH30275.1 | |||
| BC041868.1 | AAH41868.1 | |||
| CR749701.1 | CAH18479.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521717 | Chr.2:63876998 - 63959448 on Build GRCh38 | Gain |
|
| nsv582127 | Chr.2:63618350 - 64923736 on Build GRCh38 | Gain |
|
| nsv582126 | Chr.2:63346276 - 64470570 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map