Assay Details
Target Gene Details
Entrez Gene ID: | 4338 |
Gene Name: | molybdenum cofactor synthesis 2 |
Gene Aliases: |
MCBPE, MOCO1, MOCODB, MPTS |
Location: |
Chr.5:53095679-53109772 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOCS2 | NM_004531.4 | 6 | 1120 | NP_004522.1 |
| NM_176806.3 | 6 | 659 | NP_789776.1 | |
| AF091871.1 | 6 | 632 | AAD14598.1 | |
| AF117815.1 | 6 | 643 | AAD13296.1 | |
| AF155659.1 | 6 | 845 | AAF67478.1 | |
| AK126211.1 | 3 | 2176 | ||
| AK312887.1 | 6 | 645 | ||
| BC046097.1 | 6 | 806 | AAH46097.1 | |
| CD514049.1 | 6 | 655 | ||
| CR457172.1 | 4 | 429 | CAG33453.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830296 | Chr.5:52913698 - 53114423 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map