Assay Details
Target Gene Details
Entrez Gene ID: | 51574 |
Gene Name: | La ribonucleoprotein domain family member 7 |
Gene Aliases: |
ALAZS, HDCMA18P, PIP7S |
Location: |
Chr.4:112636964-112657592 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LARP7 | NM_001267039.1 | 11 | 1714 | NP_001253968.1 |
| NM_015454.2 | 9 | 1378 | NP_056269.1 | |
| NM_016648.3 | 9 | 1506 | NP_057732.2 | |
| NR_049768.1 | 9 | 1403 | ||
| AF068284.1 | 8 | 655 | AAF65503.1 | |
| AK000274.1 | 9 | 1316 | ||
| AK225900.1 | 6 | 875 | ||
| AL049996.1 | 3 | 353 | CAB43230.1 | |
| BC066945.1 | 11 | 1552 | AAH66945.1 | |
| BC107709.1 | 3 | 347 | ||
| BX647100.1 | 9 | 1240 | ||
| CA314598.1 | 5 | 798 | ||
| EU667388.1 | 9 | 1249 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525715 | Chr.4:112615590 - 112657453 on Build GRCh38 | Loss |
|
| nsv1006030 | Chr.4:112618215 - 112736658 on Build GRCh38 | Gain |
|
| nsv1014926 | Chr.4:112574087 - 112770975 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map