Assay Details
Target Gene Details
Entrez Gene ID: | 10285 |
Gene Name: | survival motor neuron domain containing 1 |
Gene Aliases: |
SMNR, SPF30, TDRD16C |
Location: |
Chr.10:110293040-110304969 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMNDC1 | NM_005871.3 | 6 | 1418 | NP_005862.1 |
| XM_005269382.3 | 7 | 1628 | XP_005269439.1 | |
| BC011234.1 | 6 | 1408 | AAH11234.1 |
More Information
Set Membership: |
|

Genomic Map