Assay Details
Target Gene Details
Entrez Gene ID: | 10962 |
Gene Name: | myeloid/lymphoid or mixed-lineage leukemia; translocated to, 11 |
Gene Aliases: |
AF1Q |
Location: |
Chr.1:151059675-151068497 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLLT11 | NM_006818.3 | 1 | 99 | NP_006809.1 |
| AK056089.1 | 1 | 94 | ||
| BC022448.1 | 1 | 371 | AAH22448.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map