Assay Details
Target Gene Details
Entrez Gene ID: | 3141 |
Gene Name: | holocarboxylase synthetase |
Gene Aliases: |
HCS |
Location: |
Chr.21:36750888-36990254 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HLCS | NM_000411.6 | 12 | 4083 | NP_000402.3 |
| NM_001242784.1 | 12 | 5017 | NP_001229713.1 | |
| NM_001242785.1 | 12 | 4176 | NP_001229714.1 | |
| XM_005260953.3 | 11 | 4045 | XP_005261010.1 | |
| XM_005260955.3 | 11 | 3916 | XP_005261012.1 | |
| XM_005260956.3 | 11 | 4092 | XP_005261013.1 | |
| XM_006723994.2 | 11 | 4030 | XP_006724057.1 | |
| XM_006723995.1 | 12 | 4208 | XP_006724058.1 | |
| XM_011529538.1 | 12 | 4063 | XP_011527840.1 | |
| XM_011529539.2 | 12 | 4068 | XP_011527841.1 | |
| XM_011529541.2 | 11 | 3919 | XP_011527843.1 | |
| XM_017028330.1 | 11 | 3866 | XP_016883819.1 | |
| D87328.1 | 12 | 4834 | BAA13332.1 |
More Information
Set Membership: |
|

Genomic Map