Assay Details
Target Gene Details
Entrez Gene ID: | 64062 |
Gene Name: | RNA binding motif protein 26 |
Gene Aliases: |
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17 |
Location: |
Chr.13:79311827-79406265 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3632816 | Chr.13:79199236 - 79375478 on Build GRCh38 | Gain |
|
| nsv832657 | Chr.13:79330843 - 79524973 on Build GRCh38 | Gain |
|
| esv3632814 | Chr.13:79199135 - 79520919 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map