Assay Details
Target Gene Details
Entrez Gene ID: | 22849 |
Gene Name: | cytoplasmic polyadenylation element binding protein 3 |
Gene Aliases: |
- |
Location: |
Chr.10:92046692-92291118 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPEB3 | NM_001178137.1 | 10 | 3987 | NP_001171608.1 |
| NM_014912.4 | 10 | 4111 | NP_055727.3 | |
| XM_005269630.4 | 10 | 4069 | XP_005269687.1 | |
| XM_006717714.3 | 11 | 4080 | XP_006717777.1 | |
| XM_006717715.3 | 11 | 4162 | XP_006717778.1 | |
| XM_011539514.2 | 10 | 4138 | XP_011537816.1 | |
| XM_011539515.2 | 11 | 4093 | XP_011537817.1 | |
| XM_011539517.2 | 9 | 4012 | XP_011537819.1 | |
| XM_011539519.2 | 8 | 3880 | XP_011537821.1 | |
| XM_011539520.2 | 8 | 3811 | XP_011537822.1 | |
| XM_017015927.1 | 7 | 2875 | XP_016871416.1 | |
| AB023157.1 | 10 | 3954 | ||
| AK127060.1 | 9 | 3059 | ||
| BC036444.2 | 10 | 4077 | AAH36444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv551923 | Chr.10:92045217 - 92076212 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map