Assay Details
Target Gene Details
Entrez Gene ID: | 81448 |
Gene Name: | olfactory receptor family 6 subfamily K member 2 |
Gene Aliases: |
OR1-17 |
Location: |
Chr.1:158699678-158700652 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6K2 | NM_001005279.1 | NP_001005279.1 | ||
| BC137022.1 | 1 | 1033 | ||
| BC137023.1 | 1 | 1074 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv547987 | Chr.1:158695182 - 158750398 on Build GRCh38 | Loss |
|
| dgv445n100 | Chr.1:158690274 - 158727028 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map