Assay Details
Target Gene Details
Entrez Gene ID: | 23001 |
Gene Name: | WD repeat and FYVE domain containing 3 |
Gene Aliases: |
ALFY, ZFYVE25 |
Location: |
Chr.4:84669537-84966391 on Build GRCh38 |
Assay Gene Location: | Within Exon 74 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WDFY3 | NM_014991.4 | 68 | 10972 | NP_055806.2 |
| XM_005262858.4 | 70 | 11013 | XP_005262915.1 | |
| XM_011531757.2 | 69 | 10924 | XP_011530059.1 | |
| XM_011531759.2 | 68 | 10886 | XP_011530061.1 | |
| XM_011531760.2 | 68 | 10824 | XP_011530062.1 | |
| XM_011531761.2 | 69 | 10879 | XP_011530063.1 | |
| XM_011531762.2 | 68 | 10873 | XP_011530064.1 | |
| XM_011531763.2 | 67 | 10770 | XP_011530065.1 | |
| XM_011531764.2 | 68 | 10840 | XP_011530066.1 | |
| XM_011531765.2 | 65 | 10575 | XP_011530067.1 | |
| XM_017007906.1 | 71 | 11141 | XP_016863395.1 | |
| XM_017007907.1 | 71 | 11138 | XP_016863396.1 | |
| XM_017007908.1 | 70 | 11070 | XP_016863397.1 | |
| XM_017007909.1 | 56 | 8766 | XP_016863398.1 | |
| AB023210.2 | 33 | 4655 | BAA76837.2 | |
| AF538685.1 | AAN15137.1 | |||
| AK055806.1 | 19 | 2334 | BAB71020.1 | |
| AK092394.1 | 8 | 1351 | ||
| BC013377.2 | 5 | 1316 | AAH13377.1 | |
| BC015214.1 | 8 | 1298 | ||
| BC065502.1 | 15 | 2322 | AAH65502.1 | |
| BC119633.2 | 5 | 1019 | ||
| BC148294.1 | 5 | 1019 | ||
| BC148298.2 | 5 | 1019 | ||
| BC150246.1 | 33 | 4676 | ||
| BX648077.1 | 25 | 3537 | ||
| BX648153.1 | 28 | 3865 |
More Information
Set Membership: |
|

Genomic Map