Assay Details
Target Gene Details
Entrez Gene ID: | 9478 |
Gene Name: | calcium binding protein 1 |
Gene Aliases: |
CALBRAIN, HCALB_BR |
Location: |
Chr.12:120640552-120680869 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CABP1 | NM_001033677.1 | NP_001028849.1 | ||
| XM_017020235.1 | XP_016875724.1 | |||
| AL533429.3 | ||||
| BC030201.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521928 | Chr.12:120603925 - 121356945 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs117649554] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map