Assay Details
Target Gene Details
Entrez Gene ID: | 148266 |
Gene Name: | zinc finger protein 569 |
Gene Aliases: |
ZAP1, ZNF |
Location: |
Chr.19:37411146-37469268 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF569 | NM_152484.2 | NP_689697.2 | ||
| XM_006723046.2 | XP_006723109.1 | |||
| XM_006723047.3 | XP_006723110.1 | |||
| XM_006723048.3 | XP_006723111.1 | |||
| XM_011526538.2 | XP_011524840.1 | |||
| XM_011526539.2 | XP_011524841.1 | |||
| XM_017026376.1 | XP_016881865.1 | |||
| XM_017026377.1 | XP_016881866.1 | |||
| XM_017026378.1 | XP_016881867.1 | |||
| XM_017026379.1 | XP_016881868.1 | |||
| XM_017026380.1 | XP_016881869.1 | |||
| XM_017026381.1 | XP_016881870.1 | |||
| XM_017026382.1 | XP_016881871.1 | |||
| AK289987.1 | ||||
| AL833408.1 | ||||
| AY835704.1 | AAV97944.1 | |||
| BC117219.1 | ||||
| CB151734.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833819 | Chr.19:37408599 - 37560697 on Build GRCh38 | Gain |
|
| esv2758759 | Chr.19:37303948 - 37560704 on Build GRCh38 | Loss |
|
| nsv579465 | Chr.19:37377650 - 37426790 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map