Assay Details
Target Gene Details
Entrez Gene ID: | 79155 |
Gene Name: | TNFAIP3 interacting protein 2 |
Gene Aliases: |
ABIN2, FLIP1, KLIP |
Location: |
Chr.4:2741660-2756376 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNIP2 | NM_001161527.1 | 2 | 173 | NP_001154999.1 |
| NM_001292016.1 | 2 | 459 | NP_001278945.1 | |
| NM_024309.3 | 2 | 459 | NP_077285.3 | |
| AF372839.1 | 2 | 459 | AAM21315.1 | |
| AJ304866.1 | 2 | 406 | CAC34835.1 | |
| AK026176.1 | 2 | 172 | BAB15382.1 | |
| AK096296.1 | 2 | 450 | ||
| BC002740.2 | 2 | 417 | AAH02740.2 | |
| BX386180.2 | 2 | 420 | ||
| DA421921.1 | 2 | 173 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470002 | Chr.4:2699371 - 2834901 on Build GRCh38 | Loss |
|
| esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
|
| nsv516797 | Chr.4:2721804 - 2834901 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map