Assay Details
Target Gene Details
Entrez Gene ID: | 4776 |
Gene Name: | nuclear factor of activated T-cells 4 |
Gene Aliases: |
NF-AT3, NF-ATC4, NFAT3 |
Location: |
Chr.14:24366911-24379604 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFATC4 | NM_001136022.2 | 10 | 2791 | NP_001129494.1 |
| NM_001198965.1 | NP_001185894.1 | |||
| NM_001198966.2 | 9 | 2433 | NP_001185895.1 | |
| NM_001198967.2 | NP_001185896.1 | |||
| NM_001288802.1 | 9 | 2582 | NP_001275731.1 | |
| NM_001320043.1 | 10 | 2791 | NP_001306972.1 | |
| NM_004554.4 | 9 | 2779 | NP_004545.2 | |
| XM_011536797.2 | 9 | 2614 | XP_011535099.1 | |
| XM_011536798.1 | 9 | 2704 | XP_011535100.1 | |
| XM_011536799.2 | XP_011535101.1 | |||
| AK293185.1 | ||||
| AK302271.1 | 9 | 2419 | ||
| AK310433.1 | 9 | 2582 | ||
| AK310445.1 | ||||
| BC008857.2 | 8 | 2280 | AAH08857.2 | |
| BC053855.1 | 9 | 2595 | AAH53855.1 | |
| EU887632.1 | 10 | 2763 | ||
| EU887633.1 | ||||
| EU887634.1 | 10 | 2763 | ||
| EU887635.1 | 10 | 2579 | ||
| EU887636.1 | ||||
| EU887637.1 | 10 | 2579 | ||
| EU887638.1 | 10 | 2632 | ||
| EU887639.1 | ||||
| EU887640.1 | 10 | 2632 | ||
| EU887641.1 | 9 | 2458 | ||
| EU887642.1 | ||||
| EU887643.1 | 9 | 2458 | ||
| EU887644.1 | 9 | 2582 | ||
| EU887645.1 | ||||
| EU887646.1 | 9 | 2582 | ||
| EU887647.1 | 8 | 2704 | ||
| EU887648.1 | ||||
| EU887649.1 | 8 | 2704 | ||
| EU887650.1 | 6 | 1455 | ||
| EU887651.1 | ||||
| EU887652.1 | 6 | 1455 | ||
| EU887653.1 | 4 | 929 | ||
| EU887654.1 | ||||
| EU887655.1 | 4 | 929 | ||
| L41066.1 | 9 | 2599 | AAA79175.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952819 | Chr.14:24367695 - 24377694 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map