Assay Details
Target Gene Details
Entrez Gene ID: | 135114 |
Gene Name: | histidine triad nucleotide binding protein 3 |
Gene Aliases: |
HINT4 |
Location: |
Chr.6:125956715-125980244 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HINT3 | NM_138571.4 | 4 | 694 | NP_612638.3 |
| AJ420577.1 | 4 | 630 | ||
| AK057688.1 | 4 | 624 | ||
| AK309857.1 | 4 | 345 | ||
| AY035387.1 | 4 | 619 | AAK71347.1 | |
| AY035388.1 | 4 | 619 | AAK71348.1 | |
| AY486460.1 | 4 | 431 | AAR89533.1 | |
| AY486461.1 | 4 | 431 | AAR89534.1 | |
| BC015732.1 | 4 | 628 | AAH15732.1 | |
| BX647228.1 | 4 | 684 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025069 | Chr.6:125973448 - 126003809 on Build GRCh38 | Loss |
|
| nsv1022677 | Chr.6:125935606 - 126195159 on Build GRCh38 | Gain |
|
| nsv1019490 | Chr.6:125927450 - 126221150 on Build GRCh38 | Loss |
|
| nsv830796 | Chr.6:125844487 - 126015022 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map