Assay Details
Target Gene Details
Entrez Gene ID: | 4692 |
Gene Name: | necdin, MAGE family member |
Gene Aliases: |
HsT16328, PWCR |
Location: |
Chr.15:23685407-23687303 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDN | NM_002487.2 | 1 | 1716 | NP_002478.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892632 | Chr.15:23375712 - 24546172 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75438249] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map