Assay Details
Target Gene Details
Entrez Gene ID: | 92346 |
Gene Name: | chromosome 1 open reading frame 105 |
Gene Aliases: |
- |
Location: |
Chr.1:172418560-172468829 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1orf105 | NM_139240.3 | 1 | 139 | NP_640333.3 |
| XM_011510152.2 | 1 | 2267 | XP_011508454.1 | |
| XM_011510153.2 | 1 | 2267 | XP_011508455.1 | |
| XM_011510154.2 | 1 | 2267 | XP_011508456.1 | |
| XM_011510155.2 | 1 | 2267 | XP_011508457.1 | |
| XM_011510156.2 | 1 | 2267 | XP_011508458.1 | |
| AL035295.1 | 1 | 139 | CAA22895.1 | |
| BC038410.1 | 1 | 137 | AAH38410.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587924 | Chr.1:172420024 - 172426974 on Build GRCh38 | Loss |
|
| esv2669467 | Chr.1:172420007 - 172427006 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map