Assay Details
Target Gene Details
Entrez Gene ID: | 112802 |
Gene Name: | keratin 71 |
Gene Aliases: |
HYPT13, K6IRS1, KRT6IRS, KRT6IRS1 |
Location: |
Chr.12:52543909-52553147 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT71 | NM_033448.2 | 9 | 2115 | NP_258259.1 |
| XM_017018749.1 | 10 | 1988 | XP_016874238.1 | |
| AJ308599.1 | 9 | 2087 | CAC43429.1 | |
| AJ308600.1 | 10 | 1994 | ||
| AJ308601.1 | 7 | 1194 | ||
| AK122795.1 | 9 | 2115 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038939 | Chr.12:52531827 - 52550411 on Build GRCh38 | Loss |
|
| nsv709 | Chr.12:52500235 - 52564462 on Build GRCh38 | Insertion |
|
| nsv558933 | Chr.12:52514689 - 52548578 on Build GRCh38 | Loss |
|
| nsv832412 | Chr.12:52390206 - 52545452 on Build GRCh38 | Loss |
|
| nsv1041903 | Chr.12:52526608 - 52550956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map