Assay Details
Target Gene Details
Entrez Gene ID: | 9798 |
Gene Name: | IST1, ESCRT-III associated factor |
Gene Aliases: |
OLC1 |
Location: |
Chr.16:71894408-71930639 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IST1 | NM_001270975.1 | NP_001257904.1 | ||
| NM_001270976.1 | NP_001257905.1 | |||
| NM_001270977.1 | NP_001257906.1 | |||
| NM_001270978.1 | NP_001257907.1 | |||
| NM_001270979.1 | NP_001257908.1 | |||
| NM_014761.3 | NP_055576.2 | |||
| AB097052.1 | BAC77405.1 | |||
| AK057902.1 | ||||
| AK291388.1 | ||||
| AK293022.1 | ||||
| AK293040.1 | ||||
| AK295066.1 | ||||
| AK297336.1 | ||||
| AK301918.1 | ||||
| BC000116.1 | AAH00116.1 | |||
| BC000430.2 | AAH00430.1 | |||
| BC004359.1 | AAH04359.1 | |||
| BC103745.1 | AAI03746.1 | |||
| BX364250.2 | ||||
| D79996.1 | BAA11491.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1055854 | Chr.16:71888241 - 72048987 on Build GRCh38 | Loss |
|
| nsv1862 | Chr.16:71900888 - 71935083 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map