Assay Details
Target Gene Details
Entrez Gene ID: | 10111 |
Gene Name: | RAD50 double strand break repair protein |
Gene Aliases: |
NBSLD, RAD502, hRad50 |
Location: |
Chr.5:132556924-132644621 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD50 | NM_005732.3 | 25 | 4190 | NP_005723.2 |
| AF057299.1 | 25 | 4110 | AAD50325.1 | |
| AF057300.1 | 25 | 4045 | AAD50326.1 | |
| BC136436.1 | 25 | 4082 | ||
| U63139.1 | 25 | 4177 | AAB07119.1 |
Target Gene Details
Entrez Gene ID: | 101927761 |
Gene Name: | T helper type 2 locus control region associated RNA |
Gene Aliases: |
TH2-LCR |
Location: |
Chr.5:132630589-132656154 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TH2LCRR | NR_132125.1 | |||
| NR_132126.1 | ||||
| AA470013.1 | ||||
| BX095327.1 |
More Information
Set Membership: |
|

Genomic Map