Assay Details
Target Gene Details
Entrez Gene ID: | 4891 |
Gene Name: | solute carrier family 11 member 2 |
Gene Aliases: |
AHMIO1, DCT1, DMT1, NRAMP2 |
Location: |
Chr.12:50952263-51028335 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC11A2 | NM_000617.2 | 16 | 3854 | NP_000608.1 |
| NM_001174125.1 | 16 | 3846 | NP_001167596.1 | |
| NM_001174126.1 | NP_001167597.1 | |||
| NM_001174127.1 | NP_001167598.1 | |||
| NM_001174128.1 | 16 | 3849 | NP_001167599.1 | |
| NM_001174129.1 | 16 | 4114 | NP_001167600.1 | |
| NM_001174130.1 | 15 | 3829 | NP_001167601.1 | |
| NR_033421.1 | 16 | 3774 | ||
| NR_033422.1 | ||||
| XM_005268911.3 | XP_005268968.1 | |||
| XM_005268912.4 | XP_005268969.1 | |||
| XM_011538404.2 | XP_011536706.1 | |||
| XM_011538405.2 | XP_011536707.1 | |||
| XM_017019355.1 | XP_016874844.1 | |||
| XM_017019356.1 | XP_016874845.1 | |||
| AB004857.1 | 16 | 3797 | BAA24933.1 | |
| AB062284.1 | BAB93467.1 | |||
| AF064484.1 | AAC21459.1 | |||
| AK128640.1 | ||||
| AL832886.1 | 6 | 3180 | ||
| BC100014.1 | 16 | 3822 | AAI00015.1 | |
| CD365137.1 | ||||
| KT455015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3549202 | Chr.12:50982946 - 50986567 on Build GRCh38 | Deletion |
|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
| esv2660448 | Chr.12:50983023 - 50986546 on Build GRCh38 | Deletion |
|
| esv3629496 | Chr.12:50982900 - 50986600 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map