Assay Details
Target Gene Details
Entrez Gene ID: | 9892 |
Gene Name: | synaptosome associated protein 91 |
Gene Aliases: |
AP180, CALM |
Location: |
Chr.6:83552885-83709408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 34 - Exon 34 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5374 | Chr.6:83542016 - 83576422 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map