Assay Details
Target Gene Details
Entrez Gene ID: | 10944 |
Gene Name: | chromosome 11 open reading frame 58 |
Gene Aliases: |
IMAGE145052, SMAP |
Location: |
Chr.11:16738601-16758354 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C11orf58 | NM_014267.5 | NP_055082.1 | ||
| XM_017017143.1 | XP_016872632.1 | |||
| AK057169.1 | ||||
| AK315382.1 | ||||
| BC007103.1 | AAH07103.1 | |||
| BC016352.1 | AAH16352.1 | |||
| BC020937.1 | AAH20937.1 | |||
| BC103747.1 | AAI03748.1 | |||
| BP210918.1 | ||||
| BP226882.1 | ||||
| BU661194.1 | ||||
| CR456882.1 | CAG33163.1 | |||
| U51678.1 | AAB99729.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv553566 | Chr.11:16307463 - 16777497 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map