Assay Details
Target Gene Details
Entrez Gene ID: | 144715 |
Gene Name: | RAD9 checkpoint clamp component B |
Gene Aliases: |
- |
Location: |
Chr.12:110501655-110534208 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAD9B | NM_001286531.1 | 11 | 1744 | NP_001273460.1 |
| NM_001286532.1 | 11 | 1766 | NP_001273461.1 | |
| NM_001286533.1 | 12 | 1922 | NP_001273462.1 | |
| NM_001286534.1 | 12 | 1842 | NP_001273463.1 | |
| NM_001286535.1 | 11 | 1773 | NP_001273464.1 | |
| NM_001286536.1 | NP_001273465.1 | |||
| NM_152442.3 | NP_689655.3 | |||
| XM_005253849.4 | 9 | 1452 | XP_005253906.1 | |
| XM_011537971.2 | XP_011536273.1 | |||
| XM_011537972.2 | 11 | 1601 | XP_011536274.1 | |
| XM_011537974.2 | 10 | 1604 | XP_011536276.1 | |
| XM_017018877.1 | 10 | 1517 | XP_016874366.1 | |
| XM_017018878.1 | 11 | 1721 | XP_016874367.1 | |
| XM_017018879.1 | 10 | 1657 | XP_016874368.1 | |
| XM_017018880.1 | 10 | 1565 | XP_016874369.1 | |
| XM_017018881.1 | XP_016874370.1 | |||
| XM_017018882.1 | 10 | 1509 | XP_016874371.1 | |
| AK058176.1 | BAB71704.1 | |||
| AK097665.1 | BAC05138.1 | |||
| AK124109.1 | 11 | 1744 | BAC85774.1 | |
| AK302512.1 | ||||
| AY297459.1 | 11 | 1649 | AAQ62859.1 | |
| BC047645.1 | 12 | 1789 | AAH47645.1 | |
| BC068031.1 |
More Information
Set Membership: |
|

Genomic Map