Assay Details
Target Gene Details
Entrez Gene ID: | 6772 |
Gene Name: | signal transducer and activator of transcription 1 |
Gene Aliases: |
CANDF7, IMD31A, IMD31B, IMD31C, ISGF-3, STAT91 |
Location: |
Chr.2:190969036-191014250 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT1 | NM_007315.3 | 25 | 4052 | NP_009330.1 |
| XM_006712718.1 | 24 | 3845 | XP_006712781.1 | |
| XM_017004783.1 | 26 | 4082 | XP_016860272.1 | |
| AI992149.1 | 1 | 147 | ||
| BU738296.1 | 1 | 275 | ||
| CR749636.1 | CAH18430.1 | |||
| EB385742.1 | ||||
| M97935.1 | 24 | 3860 | AAB64012.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999391 | Chr.2:190884309 - 191057205 on Build GRCh38 | Gain |
|
| nsv834493 | Chr.2:190946647 - 191109514 on Build GRCh38 | Gain |
|
| nsv584065 | Chr.2:190779811 - 191011175 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map