Assay Details
Target Gene Details
Entrez Gene ID: | 25939 |
Gene Name: | SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1 |
Gene Aliases: |
CHBL2, DCIP, HDDC1, MOP-5, SBBI88 |
Location: |
Chr.20:36890882-36951843 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMHD1 | NM_015474.3 | NP_056289.2 | ||
| XM_005260384.3 | XP_005260441.1 | |||
| XM_011528761.2 | XP_011527063.1 | |||
| AB013847.1 | BAB18916.1 | |||
| AB208944.1 | BAD92181.1 | |||
| AF228421.1 | AAF32407.1 | |||
| AK027811.1 | BAB55386.1 | |||
| AK304187.1 | ||||
| AK311150.1 | ||||
| AK315169.1 | ||||
| AL050267.1 | CAB43368.1 | |||
| BC036450.2 | AAH36450.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057845 | Chr.20:36855609 - 36916270 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map