Assay Details
Target Gene Details
Entrez Gene ID: | 84230 |
Gene Name: | leucine rich repeat containing 8 family member C |
Gene Aliases: |
AD158, FAD158 |
Location: |
Chr.1:89615810-89719535 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC8C | NM_032270.4 | 1 | 184 | NP_115646.2 |
| XM_011542282.2 | XP_011540584.1 | |||
| XM_017002503.1 | XP_016857992.1 | |||
| XM_017002504.1 | XP_016857993.1 | |||
| AB081509.1 | BAB86303.1 | |||
| AK127881.1 | 1 | 121 | ||
| AL699030.1 | ||||
| BC036122.1 | 1 | 197 | AAH36122.1 | |
| BC113973.1 | AAI13974.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3586765 | Chr.1:89419740 - 89647558 on Build GRCh38 | Gain |
|
| esv3586769 | Chr.1:89593477 - 89694051 on Build GRCh38 | Gain |
|
| nsv830559 | Chr.1:89535847 - 89711962 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map