Assay Details
Target Gene Details
Entrez Gene ID: | 51347 |
Gene Name: | TAO kinase 3 |
Gene Aliases: |
DPK, JIK, MAP3K18 |
Location: |
Chr.12:118149801-118372950 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAOK3 | NM_016281.3 | 21 | 4103 | NP_057365.3 |
| XM_005253897.1 | 21 | 4101 | XP_005253954.1 | |
| XM_005253898.1 | 21 | 3992 | XP_005253955.1 | |
| XM_006719445.2 | 21 | 4029 | XP_006719508.1 | |
| XM_011538437.1 | 21 | 4047 | XP_011536739.1 | |
| XM_011538439.1 | 21 | 4020 | XP_011536741.1 | |
| XM_017019408.1 | 22 | 4312 | XP_016874897.1 | |
| XM_017019409.1 | 22 | 4436 | XP_016874898.1 | |
| XM_017019410.1 | 22 | 4432 | XP_016874899.1 | |
| XM_017019411.1 | 22 | 4256 | XP_016874900.1 | |
| XM_017019412.1 | 21 | 5123 | XP_016874901.1 | |
| XM_017019413.1 | 21 | 5096 | XP_016874902.1 | |
| AA744362.1 | 1 | 297 | ||
| AF181985.1 | 21 | 3904 | AAF25817.1 | |
| AI057643.1 | 1 | 162 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581916 | Chr.12:117356518 - 118228514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map