Assay Details
Target Gene Details
Entrez Gene ID: | 7565 |
Gene Name: | zinc finger protein 17 |
Gene Aliases: |
HPF3, KOX10 |
Location: |
Chr.19:57409252-57421939 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF17 | NM_006959.2 | NP_008890.2 | ||
| XM_017027205.1 | XP_016882694.1 | |||
| AK098183.1 | BAC05252.1 | |||
| AK127948.1 | ||||
| DA476988.1 | ||||
| DA746547.1 |
More Information
Set Membership: |
|

Genomic Map