Assay Details
Target Gene Details
Entrez Gene ID: | 4130 |
Gene Name: | microtubule associated protein 1A |
Gene Aliases: |
MAP1L, MTAP1A |
Location: |
Chr.15:43510946-43531620 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP1A | NM_002373.5 | 4 | 376 | NP_002364.5 |
| XM_017022189.1 | 5 | 666 | XP_016877678.1 | |
| AF200415.2 | 2 | 108 | AAF08305.2 | |
| AK291052.1 | 4 | 348 | ||
| BC055312.1 | 4 | 345 | AAH55312.1 | |
| BC094702.1 | 4 | 335 | AAH94702.1 | |
| BP309709.1 | 4 | 376 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832988 | Chr.15:43512443 - 43684664 on Build GRCh38 | Loss |
|
| nsv515877 | Chr.15:43509826 - 43525854 on Build GRCh38 | Loss |
|
| nsv437799 | Chr.15:43491802 - 43697320 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map