Assay Details
Target Gene Details
Entrez Gene ID: | 5601 |
Gene Name: | mitogen-activated protein kinase 9 |
Gene Aliases: |
JNK-55, JNK2, JNK2A, JNK2ALPHA, JNK2B, JNK2BETA, PRKM9, SAPK, SAPK1a, p54a, p54aSAPK |
Location: |
Chr.5:180233594-180292071 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPK9 | NM_002752.4 | 12 | 1764 | NP_002743.3 |
| NM_139068.2 | 12 | 1769 | NP_620707.1 | |
| NM_139069.2 | 12 | 1769 | NP_620708.1 | |
| NM_139070.2 | 12 | 1764 | NP_620709.1 | |
| XM_005265940.4 | 13 | 1897 | XP_005265997.1 | |
| XM_006714891.2 | 13 | 1822 | XP_006714954.1 | |
| XM_017009638.1 | 13 | 1733 | XP_016865127.1 | |
| XM_017009639.1 | 14 | 1990 | XP_016865128.1 | |
| XM_017009640.1 | 13 | 1881 | XP_016865129.1 | |
| XM_017009641.1 | 13 | 1836 | XP_016865130.1 | |
| AK289638.1 | 12 | 1660 | ||
| BC032539.1 | 11 | 1554 | AAH32539.1 | |
| BQ025589.1 | 1 | 401 | ||
| L31951.1 | 11 | 1551 | AAA56831.1 | |
| U09759.1 | 11 | 1644 | AAA74740.1 | |
| U35002.1 | AAC50608.1 | |||
| U35003.1 | 11 | 1527 | AAC50609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759398 | Chr.5:180107477 - 180302944 on Build GRCh38 | Gain |
|
| nsv1033706 | Chr.5:179975769 - 180379356 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map