Assay Details
Target Gene Details
Entrez Gene ID: | 6208 |
Gene Name: | ribosomal protein S14 |
Gene Aliases: |
EMTB, S14 |
Location: |
Chr.5:150444229-150449810 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPS14 | NM_001025070.1 | NP_001020241.1 | ||
| NM_001025071.1 | NP_001020242.1 | |||
| NM_005617.3 | NP_005608.1 | |||
| XM_017009684.1 | XP_016865173.1 | |||
| AK312179.1 | ||||
| BC020515.1 | AAH20515.1 | |||
| BC091474.1 | AAH91474.1 | |||
| BM454896.1 | ||||
| BM557014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759386 | Chr.5:150445030 - 150906180 on Build GRCh38 | Loss |
|
| nsv980744 | Chr.5:150446212 - 150447914 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map