Assay Details
Target Gene Details
Entrez Gene ID: | 116843 |
Gene Name: | solute carrier family 18 member B1 |
Gene Aliases: |
C6orf192, dJ55C23.6 |
Location: |
Chr.6:132769368-132798632 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC18B1 | NM_052831.2 | 14 | 2065 | NP_439896.1 |
| XM_017010236.1 | 14 | 2256 | XP_016865725.1 | |
| AJ420537.1 | 8 | 1211 | ||
| AK124442.1 | 14 | 1944 |
More Information
Set Membership: |
|

Genomic Map