Assay Details
Target Gene Details
Entrez Gene ID: | 100500851 |
Gene Name: | microRNA 3918 |
Gene Aliases: |
mir-3918 |
Location: |
Chr.6:158764661-158764753 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR3918 | NR_037482.1 |
Target Gene Details
Entrez Gene ID: | 94120 |
Gene Name: | synaptotagmin like 3 |
Gene Aliases: |
SLP3 |
Location: |
Chr.6:158644881-158766298 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYTL3 | NM_001009991.3 | 17 | 2132 | NP_001009991.2 |
| NM_001242384.1 | 19 | 2352 | NP_001229313.1 | |
| NM_001242394.1 | 18 | 2744 | NP_001229323.1 | |
| NM_001242395.1 | 16 | 2540 | NP_001229324.1 | |
| NM_001318745.1 | 15 | 2066 | NP_001305674.1 | |
| XM_005267215.4 | 18 | 2842 | XP_005267272.1 | |
| XM_005267218.4 | 18 | 2153 | XP_005267275.1 | |
| XM_005267222.4 | 10 | 1424 | XP_005267279.1 | |
| XM_006715605.3 | 19 | 2934 | XP_006715668.1 | |
| XM_006715606.3 | 19 | 2855 | XP_006715669.1 | |
| XM_006715611.3 | 18 | 2457 | XP_006715674.1 | |
| XM_011536254.2 | 19 | 2623 | XP_011534556.1 | |
| XM_011536255.2 | 9 | 1679 | XP_011534557.1 | |
| XM_017011496.1 | 18 | 3023 | XP_016866985.1 | |
| XM_017011497.1 | 19 | 2360 | XP_016866986.1 | |
| XM_017011498.1 | 15 | 2173 | XP_016866987.1 | |
| AI674404.1 | 1 | 151 | ||
| AK055750.1 | 15 | 2065 | ||
| AK092729.1 | 14 | 1940 | ||
| AK131092.1 | 13 | 5620 | BAC85142.1 | |
| AK298712.1 | 19 | 2178 | ||
| AK304191.1 | 10 | 1609 | ||
| AL567177.3 | 1 | 114 | ||
| BC085612.1 | 16 | 2078 | AAH85612.1 | |
| BC100830.1 | AAI00831.1 | |||
| BC100831.1 | AAI00832.1 |
More Information
Set Membership: |
|

Genomic Map