Assay Details
Target Gene Details
Entrez Gene ID: | 22901 |
Gene Name: | arylsulfatase G |
Gene Aliases: |
- |
Location: |
Chr.17:68259089-68452000 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARSG | XM_006721777.3 | XP_006721840.2 | ||
| XM_011524541.2 | XP_011522843.1 | |||
| XM_011524542.2 | XP_011522844.1 | |||
| XM_011524543.2 | XP_011522845.1 | |||
| XM_011524544.2 | XP_011522846.1 | |||
| XM_017024361.1 | XP_016879850.1 | |||
| XM_017024366.1 | XP_016879855.1 | |||
| XM_017024367.1 | XP_016879856.1 |
Target Gene Details
Entrez Gene ID: | 5573 |
Gene Name: | protein kinase cAMP-dependent type I regulatory subunit alpha |
Gene Aliases: |
ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1 |
Location: |
Chr.17:68413623-68551316 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAR1A | NM_001278433.1 | NP_001265362.1 | ||
| DA108801.1 |
Target Gene Details
Entrez Gene ID: | 55062 |
Gene Name: | WD repeat domain, phosphoinositide interacting 1 |
Gene Aliases: |
ATG18, ATG18A, WIPI49 |
Location: |
Chr.17:68421281-68457524 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WIPI1 | NM_001320772.1 | 8 | 927 | NP_001307701.1 |
| NM_017983.6 | 9 | 1010 | NP_060453.3 | |
| NR_135470.1 | 9 | 1146 | ||
| NR_135471.1 | 8 | 939 | ||
| XM_017024808.1 | 7 | 2964 | XP_016880297.1 | |
| AK000917.1 | 8 | 928 | BAA91423.1 | |
| AK301830.1 | 8 | 873 | ||
| AY691424.1 | 9 | 910 | AAV80760.1 | |
| BC039867.2 | 9 | 954 | AAH39867.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833523 | Chr.17:68347182 - 68514320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map