Assay Details
Target Gene Details
Entrez Gene ID: | 9988 |
Gene Name: | cyclin D binding myb like transcription factor 1 |
Gene Aliases: |
DMP1, DMTF, MRUL, hDMP1 |
Location: |
Chr.7:87152361-87196332 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527889 | Chr.7:86866726 - 87203287 on Build GRCh38 | Gain |
|
| nsv1016761 | Chr.7:87129228 - 87284836 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map