Assay Details
Target Gene Details
Entrez Gene ID: | 339345 |
Gene Name: | nanos C2HC-type zinc finger 2 |
Gene Aliases: |
NOS2, ZC2HC12B |
Location: |
Chr.19:45913214-45914778 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NANOS2 | NM_001029861.2 | 1 | 1195 | NP_001025032.1 |
| BC042883.1 | 1 | 1197 | AAH42883.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510770 | Chr.19:45841909 - 45921434 on Build GRCh38 | Deletion |
|
| nsv2507 | Chr.19:45895584 - 45930026 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map