Assay Details
Target Gene Details
Entrez Gene ID: | 10213 |
Gene Name: | proteasome 26S subunit, non-ATPase 14 |
Gene Aliases: |
PAD1, POH1, RPN11 |
Location: |
Chr.2:161308275-161411717 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMD14 | NM_005805.5 | NP_005796.1 | ||
| AK055128.1 | ||||
| BC066336.1 | AAH66336.1 | |||
| DA133201.1 | ||||
| U86782.1 | AAC51866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583500 | Chr.2:161203306 - 161389321 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map