Assay Details
Target Gene Details
Entrez Gene ID: | 388335 |
Gene Name: | transmembrane protein 220 |
Gene Aliases: |
- |
Location: |
Chr.17:10711084-10730329 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM220 | NM_001004313.1 | 6 | 1369 | NP_001004313.1 |
| XM_005256634.4 | 5 | 1304 | XP_005256691.1 | |
| XM_011523835.2 | 5 | 853 | XP_011522137.1 | |
| XM_011523836.2 | 4 | 1245 | XP_011522138.1 | |
| XM_017024616.1 | XP_016880105.1 | |||
| XM_017024617.1 | XP_016880106.1 | |||
| AK289624.1 | 6 | 1025 | ||
| AK295928.1 | 5 | 1034 | ||
| AY550194.1 | 6 | 1369 | AAS57719.1 | |
| BC136838.1 | 6 | 935 | ||
| BC136840.1 | 6 | 935 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2675560 | Chr.17:10710661 - 10748164 on Build GRCh38 | Deletion |
|
| nsv1065819 | Chr.17:10622619 - 10784575 on Build GRCh38 | Gain |
|
| esv3639941 | Chr.17:10710674 - 10748133 on Build GRCh38 | Loss |
|
| nsv1063900 | Chr.17:10662646 - 10729182 on Build GRCh38 | Gain |
|
| nsv833367 | Chr.17:10683841 - 10826633 on Build GRCh38 | Loss |
|
| nsv1058835 | Chr.17:10322963 - 10939602 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73974829] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map