Assay Details
Target Gene Details
Entrez Gene ID: | 78988 |
Gene Name: | mitochondrial ribosomal protein L57 |
Gene Aliases: |
MRP63, bMRP63 |
Location: |
Chr.13:21176233-21179084 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL57 | NM_024026.4 | 2 | 588 | NP_076931.1 |
| XM_017020740.1 | 2 | 222 | XP_016876229.1 | |
| AB049957.1 | 1 | 110 | BAB41010.1 | |
| BC023616.2 | 2 | 144 | AAH23616.1 | |
| BC068492.1 | 2 | 154 | AAH68492.1 | |
| BG773624.1 | 2 | 177 | ||
| BG778041.1 | 2 | 160 | ||
| BI601227.1 | 2 | 130 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1037082 | Chr.13:20932196 - 21204957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map