Assay Details
Target Gene Details
Entrez Gene ID: | 3052 |
Gene Name: | holocytochrome c synthase |
Gene Aliases: |
CCHL, LSDMCA1, MCOPS7, MLS |
Location: |
Chr.X:11111286-11123086 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HCCS | NM_001122608.2 | NP_001116080.1 | ||
| NM_001171991.2 | NP_001165462.1 | |||
| NM_005333.4 | NP_005324.3 | |||
| AK097815.1 | ||||
| AU310864.1 | ||||
| BC001691.2 | AAH01691.1 | |||
| BC095455.1 | AAH95455.1 | |||
| CR749578.1 | CAH18370.1 | |||
| DB253108.1 | ||||
| U36787.1 | AAB19007.1 |
More Information
Set Membership: |
|

Genomic Map