Assay Details
Target Gene Details
Entrez Gene ID: | 163175 |
Gene Name: | leucine rich repeat LGI family member 4 |
Gene Aliases: |
LGIL3 |
Location: |
Chr.19:35124513-35135274 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LGI4 | NM_139284.2 | NP_644813.1 | ||
| XM_011526595.1 | XP_011524897.1 | |||
| XM_017026428.1 | XP_016881917.1 | |||
| XM_017026429.1 | XP_016881918.1 | |||
| XM_017026430.1 | XP_016881919.1 | |||
| AF467954.1 | AAM49552.1 | |||
| AJ487519.1 | CAD31787.1 | |||
| AJ487959.1 | CAD32306.1 | |||
| AL832968.1 | CAH56336.1 | |||
| AY358121.1 | AAQ88488.1 | |||
| BC087848.1 | AAH87848.1 | |||
| BC136694.1 | ||||
| BC136697.1 | ||||
| BC140008.1 |
More Information
Set Membership: |
|

Genomic Map