Assay Details
Target Gene Details
Entrez Gene ID: | 64327 |
Gene Name: | limb development membrane protein 1 |
Gene Aliases: |
ACHP, C7orf2, DIF14, LSS, PPD2, THYP, TPT, ZRS |
Location: |
Chr.7:156673354-156893208 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMBR1 | NM_022458.3 | 16 | 1559 | NP_071903.2 |
| XM_005249555.2 | XP_005249612.1 | |||
| XM_005249558.2 | XP_005249615.1 | |||
| XM_017012507.1 | 17 | 1657 | XP_016867996.1 | |
| XM_017012508.1 | 17 | 1615 | XP_016867997.1 | |
| XM_017012509.1 | XP_016867998.1 | |||
| XM_017012510.1 | 15 | 1261 | XP_016867999.1 | |
| XM_017012511.1 | 15 | 1390 | XP_016868000.1 | |
| XM_017012512.1 | 15 | 1321 | XP_016868001.1 | |
| XM_017012513.1 | 16 | 1339 | XP_016868002.1 | |
| XM_017012514.1 | 17 | 1628 | XP_016868003.1 | |
| XM_017012515.1 | XP_016868004.1 | |||
| XM_017012516.1 | XP_016868005.1 | |||
| XM_017012517.1 | 17 | 1612 | XP_016868006.1 | |
| XM_017012518.1 | 18 | 1710 | XP_016868007.1 | |
| AF348513.1 | 3 | 279 | AAK31345.1 | |
| AF402318.1 | 16 | 1370 | AAK94061.1 | |
| AK021727.1 | BAB13880.1 | |||
| AK026940.1 | 3 | 285 | BAB15595.1 | |
| AK291037.1 | 16 | 1530 | ||
| AK299753.1 | ||||
| AL833934.1 | CAD38789.1 | |||
| AL834394.1 | 17 | 1461 | CAD39056.1 | |
| BC017663.1 | 16 | 1498 | AAH17663.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv609209 | Chr.7:156663585 - 156716626 on Build GRCh38 | Gain |
|
| nsv831198 | Chr.7:156629947 - 156829662 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map