Assay Details
Target Gene Details
Entrez Gene ID: | 2352 |
Gene Name: | folate receptor 3 |
Gene Aliases: |
FR-G, FR-gamma, gamma-hFR |
Location: |
Chr.11:72135710-72139892 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOLR3 | NM_000804.3 | NP_000795.2 | ||
| NM_001318045.1 | NP_001304974.1 | |||
| BC030285.1 | ||||
| BI836723.1 | ||||
| U08470.1 | AAA18381.1 | |||
| U08471.1 | AAA18382.1 | |||
| Z32564.1 | CAA83553.1 | |||
| Z32633.1 | CAA83566.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2744761 | Chr.11:72135397 - 72181194 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map