Assay Details
Target Gene Details
Entrez Gene ID: | 79003 |
Gene Name: | MIS12, kinetochore complex component |
Gene Aliases: |
2510025F08Rik, KNTC2AP, MTW1, hMis12 |
Location: |
Chr.17:5486374-5490814 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIS12 | NM_001258217.1 | 3 | 1088 | NP_001245146.1 |
| NM_001258218.1 | 2 | 694 | NP_001245147.1 | |
| NM_001258219.1 | 3 | 824 | NP_001245148.1 | |
| NM_001258220.1 | 2 | 430 | NP_001245149.1 | |
| NM_024039.2 | 3 | 930 | NP_076944.1 | |
| XM_005256797.2 | 3 | 1252 | XP_005256854.1 | |
| XM_017025033.1 | 2 | 2108 | XP_016880522.1 | |
| XM_017025034.1 | 3 | 1607 | XP_016880523.1 | |
| XM_017025035.1 | 4 | 1098 | XP_016880524.1 | |
| AK056072.1 | 2 | 672 | ||
| AK289939.1 | 3 | 930 | ||
| AL136906.1 | 3 | 896 | CAB66840.1 | |
| BC000229.1 | 2 | 480 | AAH00229.1 | |
| CR533460.1 | 1 | 343 | CAG38491.1 | |
| DA964146.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1956 | Chr.17:5470460 - 5509582 on Build GRCh38 | Deletion |
|
| nsv528302 | Chr.17:5482078 - 5493377 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map