Assay Details
Target Gene Details
Entrez Gene ID: | 1821 |
Gene Name: | dystrophin related protein 2 |
Gene Aliases: |
DRP-2 |
Location: |
Chr.X:101219786-101264497 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRP2 | NM_001171184.1 | 5 | 692 | NP_001164655.1 |
| NM_001939.2 | 7 | 975 | NP_001930.2 | |
| XM_017029333.1 | 7 | 1133 | XP_016884822.1 | |
| XM_017029334.1 | 7 | 1133 | XP_016884823.1 | |
| AB209305.1 | BAD92542.1 | |||
| AK289825.1 | 7 | 863 | ||
| AK295843.1 | 5 | 561 | ||
| BC111695.1 | 7 | 812 | ||
| U43519.1 | 7 | 924 | AAC50538.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524411 | Chr.X:101190975 - 101377649 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map