Assay Details
Target Gene Details
Entrez Gene ID: | 115701 |
Gene Name: | alpha kinase 2 |
Gene Aliases: |
HAK |
Location: |
Chr.18:58481247-58629192 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALPK2 | NM_052947.3 | 11 | 6379 | NP_443179.3 |
| XM_011525801.2 | 11 | 6612 | XP_011524103.1 | |
| XM_011525802.2 | 9 | 6999 | XP_011524104.1 | |
| XM_017025535.1 | 11 | 6613 | XP_016881024.1 | |
| XM_017025536.1 | 11 | 6613 | XP_016881025.1 | |
| XM_017025537.1 | XP_016881026.1 | |||
| XM_017025538.1 | 8 | 5497 | XP_016881027.1 | |
| XM_017025539.1 | 8 | 4245 | XP_016881028.1 | |
| AL832018.2 | 8 | 4711 | CAD89922.1 | |
| AY044450.1 | 8 | 4446 | AAK95952.1 | |
| BX647446.1 | 7 | 3857 | ||
| BX647639.1 | 8 | 5245 | ||
| BX647796.1 | 11 | 6366 |
More Information
Set Membership: |
|

Genomic Map