Assay Details
Target Gene Details
Entrez Gene ID: | 3615 |
Gene Name: | inosine monophosphate dehydrogenase 2 |
Gene Aliases: |
IMPD2, IMPDH-II |
Location: |
Chr.3:49024325-49029750 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IMPDH2 | NM_000884.2 | NP_000875.2 | ||
| XM_006713128.3 | XP_006713191.1 | |||
| XM_017006349.1 | XP_016861838.1 | |||
| XM_017006350.1 | XP_016861839.1 | |||
| AY545560.1 | AAS50155.1 | |||
| BC006124.1 | AAH06124.1 | |||
| BC012840.1 | AAH12840.1 | |||
| BC015567.2 | AAH15567.1 | |||
| J04208.1 | AAA36112.1 |
More Information
Set Membership: |
|

Genomic Map